missing translation for 'onlineSavingsMsg'
Learn More

Human CLEC12B Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Product Code. 18734454
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18734454 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18734454 Supplier R&D Systems Supplier No. FAB11476R100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLEC12B Monoclonal antibody specifically detects CLEC12B in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC12B
Applications Flow Cytometry
Classification Monoclonal
Clone 1062714
Conjugate Alexa Fluor 647
Dilution Flow Cytometry
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q2HXU8
Gene Alias C-type lectin domain family 12 member B, C-type lectin domain family 12, member B, Macrophage Antigen H, UNQ5782, UNQ5782/PRO16089
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human CLEC12B, Leu65-Asp276, Accession # Q2HXU8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387837
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.