missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CISD2 Full-length ORF (NP_001008389.1, 1 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00493856-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100764

  • 508.00€ / 25µg

Estimated Shipment: 27-04-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a zinc finger protein that localizes to the endoplasmic reticulum. The encoded protein binds an iron/sulfur cluster and may be involved in calcium homeostasis. Defects in this gene are a cause of Wolfram syndrome 2 (WFS2). [provided by RefSeq]

Sequence: MVLESVARIVKVQLPAYLKRLPVPESITGFARLTVSEWLRLLPFLGVLALLGYLAVRPFLPKKKQQKDSLINLKIQKENPKVVNEINIEDLCLTKAAYCRCWRSKTFPACDGSHNKHNELTGDNVGPLILKKKEV
Specifications

Specifications

NP_001008389.1
Liquid
493856
CISD2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVLESVARIVKVQLPAYLKRLPVPESITGFARLTVSEWLRLLPFLGVLALLGYLAVRPFLPKKKQQKDSLINLKIQKENPKVVNEINIEDLCLTKAAYCRCWRSKTFPACDGSHNKHNELTGDNVGPLILKKKEV
RUO
CISD2
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ERIS/Miner1/WFS2/ZCD2
CISD2
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers