missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CISD2 Full-length ORF (NP_001008389.1, 1 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001008389.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 493856 |
Molecular Weight (g/mol) | 41.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100764
|
Abnova™
H00493856-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16190754
|
Abnova™
H00493856-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a zinc finger protein that localizes to the endoplasmic reticulum. The encoded protein binds an iron/sulfur cluster and may be involved in calcium homeostasis. Defects in this gene are a cause of Wolfram syndrome 2 (WFS2). [provided by RefSeq]
Sequence: MVLESVARIVKVQLPAYLKRLPVPESITGFARLTVSEWLRLLPFLGVLALLGYLAVRPFLPKKKQQKDSLINLKIQKENPKVVNEINIEDLCLTKAAYCRCWRSKTFPACDGSHNKHNELTGDNVGPLILKKKEVSpecifications
NP_001008389.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
41.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVLESVARIVKVQLPAYLKRLPVPESITGFARLTVSEWLRLLPFLGVLALLGYLAVRPFLPKKKQQKDSLINLKIQKENPKVVNEINIEDLCLTKAAYCRCWRSKTFPACDGSHNKHNELTGDNVGPLILKKKEV | |
RUO | |
CISD2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
493856 | |
CISD2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERIS/Miner1/WFS2/ZCD2 | |
CISD2 | |
Recombinant | |
wheat germ expression system |