Learn More
Human CFL1 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11669-100
This item is not returnable.
View return policy
Description
Cofilin Monoclonal antibody specifically detects Cofilin in Human samples. It is validated for Simple Western,Western Blot,Immunohistochemistry,Immunocytochemistry
Specifications
| Cofilin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| 18 kDa phosphoprotein, CFL, cofilin, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, HEL-S-15, p18 | |
| Synthetic Peptide, Accession # P23528 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Simple Western, Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 1093310 | |
| Western Blot 0.5 μg/mL, Simple Western 10-100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| P23528 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1072 | |
| Human | |
| Antibody | |
| IgG2b |