missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CENPL Full-length ORF (AAH33154.1, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00091687-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16116643

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

CENPL is a subunit of a CENPH (MIM 605607)-CENPI (MIM 300065)-associated centromeric complex that targets CENPA (MIM 117139) to centromeres and is required for proper kinetochore function and mitotic progression (Okada et al., 2006) [PubMed 16622420].[supplied by OMIM]

Sequence: MDSYSAPESTPSASSRPEDYFIGATPLQKRLESVRKQSSFILTPPRRKIPQCSQLQEDVDPQKVAFLLHKQWTLYSLTPLYKFSYSNLKEYSRLLNAFIVAEKQKGLAVEVGEDFNIKVIFSTLLGMKGTQRDPEAFLVQGLILSPRLEYSGTILVDCNLCLLGSSDPSTLAFQVAGTAGACHHTRIVSKSQLPSENREGKVLWTGWFCCVFGDSLLETVSEDFTCLPLFLANGAESNTAIIGTWFQKTFDCYFSPLAINAFNLSWMAAMWTACKMDHYVATTEFLWSVPCSPQSLDISFAIHPEDAKALWDSVHKTPGEVTQEEVDLFMDCLYSHFHRHFKIHLSATRLVRVSTSVASAHTDGKIKILCHKYLIGVLAYLTELAIFQIE
Specifications

Specifications

AAH33154.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1orf155/CENP-L/FLJ31044/FLJ31786/RP3-383J4.1/dJ383J4.3
CENPL
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
91687
CENPL (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDSYSAPESTPSASSRPEDYFIGATPLQKRLESVRKQSSFILTPPRRKIPQCSQLQEDVDPQKVAFLLHKQWTLYSLTPLYKFSYSNLKEYSRLLNAFIVAEKQKGLAVEVGEDFNIKVIFSTLLGMKGTQRDPEAFLVQGLILSPRLEYSGTILVDCNLCLLGSSDPSTLAFQVAGTAGACHHTRIVSKSQLPSENREGKVLWTGWFCCVFGDSLLETVSEDFTCLPLFLANGAESNTAIIGTWFQKTFDCYFSPLAINAFNLSWMAAMWTACKMDHYVATTEFLWSVPCSPQSLDISFAIHPEDAKALWDSVHKTPGEVTQEEVDLFMDCLYSHFHRHFKIHLSATRLVRVSTSVASAHTDGKIKILCHKYLIGVLAYLTELAIFQIE
RUO
CENPL
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers