missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CENPL Full-length ORF (AAH33154.1, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH33154.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 91687 |
Molecular Weight (g/mol) | 70.1kDa |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
16116643
|
Abnova™
H00091687-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16126643
|
Abnova™
H00091687-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Descripción
CENPL is a subunit of a CENPH (MIM 605607)-CENPI (MIM 300065)-associated centromeric complex that targets CENPA (MIM 117139) to centromeres and is required for proper kinetochore function and mitotic progression (Okada et al., 2006) [PubMed 16622420].[supplied by OMIM]
Sequence: MDSYSAPESTPSASSRPEDYFIGATPLQKRLESVRKQSSFILTPPRRKIPQCSQLQEDVDPQKVAFLLHKQWTLYSLTPLYKFSYSNLKEYSRLLNAFIVAEKQKGLAVEVGEDFNIKVIFSTLLGMKGTQRDPEAFLVQGLILSPRLEYSGTILVDCNLCLLGSSDPSTLAFQVAGTAGACHHTRIVSKSQLPSENREGKVLWTGWFCCVFGDSLLETVSEDFTCLPLFLANGAESNTAIIGTWFQKTFDCYFSPLAINAFNLSWMAAMWTACKMDHYVATTEFLWSVPCSPQSLDISFAIHPEDAKALWDSVHKTPGEVTQEEVDLFMDCLYSHFHRHFKIHLSATRLVRVSTSVASAHTDGKIKILCHKYLIGVLAYLTELAIFQIEEspecificaciones
AAH33154.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDSYSAPESTPSASSRPEDYFIGATPLQKRLESVRKQSSFILTPPRRKIPQCSQLQEDVDPQKVAFLLHKQWTLYSLTPLYKFSYSNLKEYSRLLNAFIVAEKQKGLAVEVGEDFNIKVIFSTLLGMKGTQRDPEAFLVQGLILSPRLEYSGTILVDCNLCLLGSSDPSTLAFQVAGTAGACHHTRIVSKSQLPSENREGKVLWTGWFCCVFGDSLLETVSEDFTCLPLFLANGAESNTAIIGTWFQKTFDCYFSPLAINAFNLSWMAAMWTACKMDHYVATTEFLWSVPCSPQSLDISFAIHPEDAKALWDSVHKTPGEVTQEEVDLFMDCLYSHFHRHFKIHLSATRLVRVSTSVASAHTDGKIKILCHKYLIGVLAYLTELAIFQIE | |
RUO | |
CENPL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
91687 | |
CENPL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C1orf155/CENP-L/FLJ31044/FLJ31786/RP3-383J4.1/dJ383J4.3 | |
CENPL | |
Recombinant | |
wheat germ expression system |