Learn More
Human CEACAM-16 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand:  R&D Systems FAB9885U-100UG
Product Code. 18714477
  • 467.00€ / 100µg
Description
CEACAM16 Monoclonal antibody specifically detects CEACAM16 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEACAM16
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
carcinoembryonic antigen-related cell adhesion molecule 16, CEAL2, DFNA4B
Chinese Hamster Ovary cell line CHO-derived human CEACAM-16, Met1-Gly425, Accession # Q2WEN9
100 μg
Primary
Human
Purified
Flow Cytometry
2747B
Flow Cytometry
Q2WEN9
Rabbit
Protein A or G purified from cell culture supernatant
RUO
388551
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.