missing translation for 'onlineSavingsMsg'
Learn More

Human CD39/ENTPD1 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

214.00€ - 503.00€

Specifications

Antigen CD39/ENTPD1
Clone 2991A
Dilution Flow Cytometry 0.25 μg/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30241007
View Documents
R&D Systems
MAB11582-100
100 μg
503.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30241016
View Documents
R&D Systems
MAB11582-SP
25 μg
214.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD39/ENTPD1 Monoclonal antibody specifically detects CD39/ENTPD1 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD39/ENTPD1
Flow Cytometry 0.25 μg/10^6 cells
Monoclonal
Purified
RUO
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
ADPase, ATPDase, ATP-diphosphatase, CD39, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, ENTPD1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPD1, NTPDase 1, NTPDase-1
Chinese Hamster Ovary cell line, CHO-derived human CD39/ENTPD1, Thr38-Val478, Accession # P49961
Primary
Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
2991A
Flow Cytometry
Unconjugated
Rabbit
Human
P49961
953
IgG
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.