missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CD25 Carrier-Free

Product Code. p-7091777
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 15558826

Brand: Invitrogen™ 34870280

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Recombinant Protein

CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency. Description: The human soluble IL-2 receptor alpha chain (sIL-2Ra) is generated by proteolytic cleavage of the cell-surface bound receptor on activated T cells. The IL-2Ra (also known as CD25) is expressed by early progenitors of the T and B lineage, as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 with low affinity. However, association with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) results in formation of the high affinity IL-2R. CD25 plays a role in lymphocyte differentiation and activation/proliferation. sIL-2Ra has been detected in the serum of patients with autoimmune disease and leukemia. Reports have demonstrated that addition of sIL-2Ra can inhibit IL-2-mediated signaling and enhance Th17 responses. Applications Reported: Recombinant human soluble IL-2Ra is biologically active and can enhance proliferation of mouse CTLL-2 cells stimulated with 1ng/mL IL-2. Applications Tested: The ED50 of this protein, as determined by increased proliferation of mouse CTLL-2 cells stimulated with 1ng/mL IL-2, is 0.5-1.5ug/mL. Source: Insect cells (amino acids Glu22-Gln240; accession &Num;: NP_000408.1). Bioactivity: The ED50 of this protein, as determined by increased proliferation of mouse CTLL-2 cells stimulated with 1ng/mL IL-2, is 0.5-1.5ug/mL. Endotoxin: Less than 0.1ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE and HPLC. Molecular Weight: 24.8 kDa. Storage and handling: For best recovery, quick spin vial prior to opening. Reconstitute in 1X PBS, pH 7.2 to a concentration of 0.1-1.0mg/mL. Do not vortex. Use in sterile environment. Store lyophilized protein at less than or equal to -20°C. Reconstituted protein solution can be stored with carrier-protein (e.g., 0.1% BSA) at less than or equal to -20°C.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01589
For Use With (Application) Control, Functional Assay
Formulation PBS with no preservative
Gene ID (Entrez) 3559
Molecular Weight (g/mol) 24.8kDa
Name Human CD25 Carrier-Free
Purification Method SDS-PAGE and HPLC
Quantity 25 μg
Source Insect Cells
Storage Requirements -20°C
Regulatory Status RUO
Structural Form Insect cells (amino acids Glu22- Gln240; Accession #: NP_000408.1)
Endotoxin Concentration Less than 0.1ng/μg cytokine as determined by the LAL assay
Common Name Human CD25 Carrier-Free
Gene Symbol IL2RA
Biological Activity The ED50 of this protein, as determined by increased proliferation of mouse CTLL-2 cells stimulated with 1ng/mL IL-2, is 0.5-1.5μg/mL.
Product Type Recombinant Protein, Carrier-Free
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species Baculovirus
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade Affinity chromatography
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.