Learn More
Human CD19 Antibody, R&D Systems™
Mouse Monoclonal Antibody
244.00€ - 504.00€
Specifications
| Antigen | CD19 |
|---|---|
| Clone | 1083805 |
| Dilution | Flow Cytometry 0.25 μg/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30243663
|
R&D Systems
MAB11581-SP |
25 μg |
244.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30243671
|
R&D Systems
MAB11581-100 |
100 μg |
504.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow CytometrySpecifications
| CD19 | |
| Flow Cytometry 0.25 μg/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 | |
| Whole Cells, Accession # P15391 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1083805 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P15391 | |
| 930 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |