missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB48671-SP
This item is not returnable.
View return policy
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry
Specifications
| CD19 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 | |
| Chinese Hamster Ovary cell line, CHO-derived human CD19, Met1-Lys291, Accession # P15391 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry, Immunocytochemistry | |
| 1062947 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P15391 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 930 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction