Learn More
Human CCL23/MPIF-1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF371-SP
This item is not returnable.
View return policy
Description
CCL23/MPIF-1 Polyclonal specifically detects CCL23/MPIF-1 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL23/MPIF-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCL23 | |
| E. coli-derived recombinant human CCL23/MPIF-1 Arg22-Asn120 Accession # P55773 | |
| 25 μg | |
| Primary | |
| Detects human CCL23/MPIF-1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) MIP-1 delta , rhHCC-1, rhMIP-1 alpha and recombinant mouse MIP-1 gamma is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.5-2.5 ug/mL | |
| P55773 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6368 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |