Learn More
Human CCL23/MPIF-1 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF371-SP
Product Code. 18750119
  • 196.00€ / 25µg
Description
CCL23/MPIF-1 Polyclonal specifically detects CCL23/MPIF-1 in Human samples. It is validated for Western Blot, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL23/MPIF-1
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CCL23
E. coli-derived recombinant human CCL23/MPIF-1 Arg22-Asn120 Accession # P55773
25 μg
Primary
Detects human CCL23/MPIF-1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) MIP-1 delta , rhHCC-1, rhMIP-1 alpha and recombinant mouse MIP-1 gamma is observed.
Human
IgG
Western Blot, Neutralization
LYOPH
Western Blot 0.1 ug/mL, Neutralization 0.5-2.5 ug/mL
P55773
Goat
Affinity Purified
RUO
6368
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.