missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL11/Eotaxin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems MAB320-500
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
Spezifikation
| CCL11/Eotaxin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, Eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6356 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Neutralization, ELISA | |
| 43911 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 1-5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P51671 | |
| CCL11 | |
| E. coli-derived recombinant human CCL11/Eotaxin Gly24-Pro97 Accession # P51671 | |
| 500 μg | |
| Primary | |
| Detects human CCL11/Eotaxin in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 9/10/MIP-1 gamma , 14, 17, 19, 20, 21, 25, recombinant mouse CCL2, 3 4 56 7, 9/10/MIP-1 gamma , 11, 21, or 25. | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur