missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCDC87 Full-length ORF (ENSP00000328487, 1 a.a. - 849 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16141313
Change view
Click to view available options
Quantity:
2 μg
Unit Size:
2µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16141313 2 μg 2µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16141313 Supplier Abnova™ Supplier No. H00055231P01.2ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Sequence: MMEPPKPEPELQRFYHRLLRPLSLFPTRTTSPEPQKRPPQEGRILQSFPLAKLTVASLCSQVAKLLAGSGIAAGVPPEARLRLIKVILDELKCSWREPPAELSLSHKNNQKLRKRLEAYVLLSSEQLFLRYLHLLVTMSTPRGVFTESATLTRLAASLARDCTLFLTSPNVYRGLLADFQALLRAEQASGDVDKLHPVCPAGTFKLCPIPWPHSTGFAQVQCSNLNLNYLIQLSRPPEFLNEPGRMDPVKELKSIPRLKRKKPFHWLPSIGKKREIDISSSQMVSLPSYPVAPTSRASPSPFCPELRRGQSMPSLREGWRLADELGLPPLPSRPLTPLVLATESKPELTGLIVAEDLKQLIKKMKLEGTRYPPLDSGLPPLLGVVTRHPAAGHRLEELEKMLRNLQEEEASGQWDPQPPKSFPLHPQPVTITLKLRNEVVVQAAAVRVSDRNFLDSFHIEGAGALYNHLAGELDPKAIEKMDIDNFVGSTTREVYKELMSHVSSDHLHFDQGPLVEPAADKDWSTFLSSAFLRQEKQPQIINPELVGLYSQRANTLQSNTKKMPSLPSLQATKSWEKWSNKASLMNSWKTTLSVDDYFKYLTNHETDFLHVIFQMHEEEVPVEIVAPARESLEIQHPPPLLEDEEPDFVPGEWDWNTVLEHRLGAGKTPHLGEPHKILSLQKHLEQLWSVLEVPDKDQVDMTIKYSSKARLRQLPSLVNAWERALKPIQLREALLARLEWFEGQASNPNRFFKKTNLSSSHFLEENQVRSHLHRKLNLMESSLVSLLEEIELIFGEPVIFKGRPYLDKMKSDKVEMLYWLQQQRRVRHLVSALKDPHQSTLFRSSAASL

Specifications

Accession Number ENSP00000328487
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55231
Molecular Weight (g/mol) 122.8kDa
Name CCDC87 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MMEPPKPEPELQRFYHRLLRPLSLFPTRTTSPEPQKRPPQEGRILQSFPLAKLTVASLCSQVAKLLAGSGIAAGVPPEARLRLIKVILDELKCSWREPPAELSLSHKNNQKLRKRLEAYVLLSSEQLFLRYLHLLVTMSTPRGVFTESATLTRLAASLARDCTLFLTSPNVYRGLLADFQALLRAEQASGDVDKLHPVCPAGTFKLCPIPWPHSTGFAQVQCSNLNLNYLIQLSRPPEFLNEPGRMDPVKELKSIPRLKRKKPFHWLPSIGKKREIDISSSQMVSLPSYPVAPTSRASPSPFCPELRRGQSMPSLREGWRLADELGLPPLPSRPLTPLVLATESKPELTGLIVAEDLKQLIKKMKLEGTRYPPLDSGLPPLLGVVTRHPAAGHRLEELEKMLRNLQEEEASGQWDPQPPKSFPLHPQPVTITLKLRNEVVVQAAAVRVSDRNFLDSFHIEGAGALYNHLAGELDPKAIEKMDIDNFVGSTTREVYKELMSHVSSDHLHFDQGPLVEPAADKDWSTFLSSAFLRQEKQPQIINPELVGLYSQRANTLQSNTKKMPSLPSLQATKSWEKWSNKASLMNSWKTTLSVDDYFKYLTNHETDFLHVIFQMHEEEVPVEIVAPARESLEIQHPPPLLEDEEPDFVPGEWDWNTVLEHRLGAGKTPHLGEPHKILSLQKHLEQLWSVLEVPDKDQVDMTIKYSSKARLRQLPSLVNAWERALKPIQLREALLARLEWFEGQASNPNRFFKKTNLSSSHFLEENQVRSHLHRKLNLMESSLVSLLEEIELIFGEPVIFKGRPYLDKMKSDKVEMLYWLQQQRRVRHLVSALKDPHQSTLFRSSAASL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ10786
Common Name CCDC87
Gene Symbol CCDC87
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.