Learn More
Abnova™ Human CATSPER1 Partial ORF (NP_444282.2, 681 a.a. - 780 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00117144-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Calcium ions play a primary role in the regulation of sperm motility. This gene belongs to a family of putative cation channels that are specific to spermatozoa and localize to the flagellum. The protein family features a single repeat with six membrane-spanning segments and a predicted calcium-selective pore region. [provided by RefSeq]
Sequence: GLEKAKQERAARIQEKLLEDSLTELRAAEPKEVASEGTMLKRLIEKKFGTMTEKQQELLFHYLQLVASVEQEQQKFRSQAAVIDEIVDTTFEAGEEDFRNSpecifications
NP_444282.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GLEKAKQERAARIQEKLLEDSLTELRAAEPKEVASEGTMLKRLIEKKFGTMTEKQQELLFHYLQLVASVEQEQQKFRSQAAVIDEIVDTTFEAGEEDFRN | |
RUO | |
CATSPER1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
117144 | |
CATSPER1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CATSPER/MGC33335/MGC33368 | |
CATSPER1 | |
Recombinant | |
wheat germ expression system |