missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CATSPER1 Partial ORF (NP_444282.2, 681 a.a. - 780 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_444282.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 117144
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16101557
View Documents Promotion Details
Abnova™
H00117144-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16191547
View Documents Promotion Details
Abnova™
H00117144-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

Calcium ions play a primary role in the regulation of sperm motility. This gene belongs to a family of putative cation channels that are specific to spermatozoa and localize to the flagellum. The protein family features a single repeat with six membrane-spanning segments and a predicted calcium-selective pore region. [provided by RefSeq]

Sequence: GLEKAKQERAARIQEKLLEDSLTELRAAEPKEVASEGTMLKRLIEKKFGTMTEKQQELLFHYLQLVASVEQEQQKFRSQAAVIDEIVDTTFEAGEEDFRN
Specifications

Specifications

NP_444282.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CATSPER/MGC33335/MGC33368
CATSPER1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
117144
CATSPER1 (Human) Recombinant Protein (Q01)
GLEKAKQERAARIQEKLLEDSLTELRAAEPKEVASEGTMLKRLIEKKFGTMTEKQQELLFHYLQLVASVEQEQQKFRSQAAVIDEIVDTTFEAGEEDFRN
RUO
CATSPER1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications