missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cardiotrophin-1/CT-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF612
This item is not returnable.
View return policy
Description
Cardiotrophin-1/CT-1 Polyclonal specifically detects Cardiotrophin-1/CT-1 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| Cardiotrophin-1/CT-1 | |
| Polyclonal | |
| Unconjugated | |
| Q16619 | |
| CTF1 | |
| E. coli-derived recombinant human Cardiotrophin-1/CT-1 Ser2-Ala201 Accession # Q16619 | |
| 100 μg | |
| Primary | |
| Detects human Cardiotrophin-1/CT-1 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Cardiotrophin-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.1-0.3 ug/mL | |
| cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1489 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction