missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cardiotrophin-1/CT-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2603-100
This item is not returnable.
View return policy
Description
Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Human samples. It is validated for ELISA.
Specifications
| Cardiotrophin-1/CT-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1 | |
| E. coli-derived recombinant human Cardiotrophin-1/CT-1 Ser2-Ala201 Accession # Q16619 | |
| 100 μg | |
| Primary | |
| Detects human Cardiotrophin-1/CT-1 in direct ELISAs. | |
| Human | |
| Purified |
| ELISA | |
| 89230 | |
| ELISA | |
| Q16619 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 1489 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction