missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cardiotrophin-1/CT-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB260-SP
This item is not returnable.
View return policy
Description
Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Human samples. It is validated for Western Blot.
Specifications
| Cardiotrophin-1/CT-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 1489 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 89228 | |
| Western Blot 1 ug/mL | |
| Q16619 | |
| CTF1 | |
| E. coli-derived recombinant human Cardiotrophin-1/CT-1 Ser2-Ala201 Accession # Q16619 | |
| 25 μg | |
| Primary | |
| Detects human Cardiotrophin-1/CT-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Cardiotrophin-1/CT-1 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction