missing translation for 'onlineSavingsMsg'
Learn More

Human Carboxylesterase 2/CES2 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18780447
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18780447 25 μg 25µg
18762107 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18780447 Supplier R&D Systems Supplier No. MAB56571SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carboxylesterase 2/CES2 Monoclonal specifically detects Carboxylesterase 2/CES2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carboxylesterase 2/CES2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 861429
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O00748
Gene Alias carboxylesterase 2 (intestine, liver), carboxylesterase 2EC 3.1.1.1, CE-2, CE-2PCE-2, CES2, CES2A1, EC 3.1.1, EC 3.1.1.84, hCE-2, iCE, intestinal carboxylesterase; liver carboxylesterase-2, PCE-2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase 2/CES2, Gln27-Leu559, Accession # O00748
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8824
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.