missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CAPN11 Full-length ORF (AAH33733.1, 1 a.a. - 702 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011131-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106222

  • 349.00€ / 10µg

Estimated Shipment: 06-07-2023
to see stock.
Add to basket

Description

Description

Calpains constitute a family of intracellular calcium-dependent cysteine proteases. There are eight members in this superfamily. They consist of a variable 80 kDa subunit and an invariant 30 kDa subunit. This calpain protein appears to have protease activity and calcium-binding ability. A similar mouse protein may play a functional role in spermatogenesis and in the regulation of calcium-dependent signal transduction events during meiosis. [provided by RefSeq]

Sequence: MVAHINNSRLKAKGVGQHDNAQNFGNQSFEELRAACLRKGELFEDPLFPAEPSSLGFKDLGPNSKNVQNISWQRPKDIINNPLFIMDGISPTDICQGILGDCWLLAAIGSLTTCPKLLYRVVPRGQSFKKNYAGIFHFQIWQFGQWVNVVVDDRLPTKNDKLVFVHSTERSEFWSALLEKAYAKLSGSYEALSGGSTMEGLEDFTGGVAQSFQLQRPPQNLLRLLRKAVERSSLMGCSIEVTSDSELESMTDKMLVRGHAYSVTGLQDVHYRGKMETLIRVRNPWGRIEWNGAWSDSAREWEEVASDIQMQLLHKTEDGEFWMSYQDFLNNFTLLEICNLTPDTLSGDYKSYWHTTFYEGSWRRGSSAGGCRNHPGTFWTNPQFKISLPEGDDPEDDAEGNVVACTCLVALMQKNWRHARQQGAQLQTIGFVLYAVPKEFQNIQDVHLKKEFFTKYQDHGFSEIFTNSREVSSQLRLPPGEYIIIPSTFEPHRDADFLLRVFTEKHSESWELDEVNYAEQLQEEKVSEDDMDQDFLHLFKIVAGEGKEIGVYELQRLLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARYADDDLIIDFDSFISCFLRLKTMFTFFLTMDPKNTGHICLSLEQWLQMTMWG
Specifications

Specifications

AAH33733.1
Liquid
11131
CAPN11 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVAHINNSRLKAKGVGQHDNAQNFGNQSFEELRAACLRKGELFEDPLFPAEPSSLGFKDLGPNSKNVQNISWQRPKDIINNPLFIMDGISPTDICQGILGDCWLLAAIGSLTTCPKLLYRVVPRGQSFKKNYAGIFHFQIWQFGQWVNVVVDDRLPTKNDKLVFVHSTERSEFWSALLEKAYAKLSGSYEALSGGSTMEGLEDFTGGVAQSFQLQRPPQNLLRLLRKAVERSSLMGCSIEVTSDSELESMTDKMLVRGHAYSVTGLQDVHYRGKMETLIRVRNPWGRIEWNGAWSDSAREWEEVASDIQMQLLHKTEDGEFWMSYQDFLNNFTLLEICNLTPDTLSGDYKSYWHTTFYEGSWRRGSSAGGCRNHPGTFWTNPQFKISLPEGDDPEDDAEGNVVACTCLVALMQKNWRHARQQGAQLQTIGFVLYAVPKEFQNIQDVHLKKEFFTKYQDHGFSEIFTNSREVSSQLRLPPGEYIIIPSTFEPHRDADFLLRVFTEKHSESWELDEVNYAEQLQEEKVSEDDMDQDFLHLFKIVAGEGKEIGVYELQRLLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARYADDDLIIDFDSFISCFLRLKTMFTFFLTMDPKNTGHICLSLEQWLQMTMWG
RUO
CAPN11
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
107kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
calpain11
CAPN11
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers