missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CAPN11 Full-length ORF (AAH33733.1, 1 a.a. - 702 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH33733.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11131 |
Molecular Weight (g/mol) | 107kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16116222
|
Abnova™
H00011131-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16106222
|
Abnova™
H00011131-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Calpains constitute a family of intracellular calcium-dependent cysteine proteases. There are eight members in this superfamily. They consist of a variable 80 kDa subunit and an invariant 30 kDa subunit. This calpain protein appears to have protease activity and calcium-binding ability. A similar mouse protein may play a functional role in spermatogenesis and in the regulation of calcium-dependent signal transduction events during meiosis. [provided by RefSeq]
Sequence: MVAHINNSRLKAKGVGQHDNAQNFGNQSFEELRAACLRKGELFEDPLFPAEPSSLGFKDLGPNSKNVQNISWQRPKDIINNPLFIMDGISPTDICQGILGDCWLLAAIGSLTTCPKLLYRVVPRGQSFKKNYAGIFHFQIWQFGQWVNVVVDDRLPTKNDKLVFVHSTERSEFWSALLEKAYAKLSGSYEALSGGSTMEGLEDFTGGVAQSFQLQRPPQNLLRLLRKAVERSSLMGCSIEVTSDSELESMTDKMLVRGHAYSVTGLQDVHYRGKMETLIRVRNPWGRIEWNGAWSDSAREWEEVASDIQMQLLHKTEDGEFWMSYQDFLNNFTLLEICNLTPDTLSGDYKSYWHTTFYEGSWRRGSSAGGCRNHPGTFWTNPQFKISLPEGDDPEDDAEGNVVACTCLVALMQKNWRHARQQGAQLQTIGFVLYAVPKEFQNIQDVHLKKEFFTKYQDHGFSEIFTNSREVSSQLRLPPGEYIIIPSTFEPHRDADFLLRVFTEKHSESWELDEVNYAEQLQEEKVSEDDMDQDFLHLFKIVAGEGKEIGVYELQRLLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARYADDDLIIDFDSFISCFLRLKTMFTFFLTMDPKNTGHICLSLEQWLQMTMWGSpecifications
AAH33733.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
107kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVAHINNSRLKAKGVGQHDNAQNFGNQSFEELRAACLRKGELFEDPLFPAEPSSLGFKDLGPNSKNVQNISWQRPKDIINNPLFIMDGISPTDICQGILGDCWLLAAIGSLTTCPKLLYRVVPRGQSFKKNYAGIFHFQIWQFGQWVNVVVDDRLPTKNDKLVFVHSTERSEFWSALLEKAYAKLSGSYEALSGGSTMEGLEDFTGGVAQSFQLQRPPQNLLRLLRKAVERSSLMGCSIEVTSDSELESMTDKMLVRGHAYSVTGLQDVHYRGKMETLIRVRNPWGRIEWNGAWSDSAREWEEVASDIQMQLLHKTEDGEFWMSYQDFLNNFTLLEICNLTPDTLSGDYKSYWHTTFYEGSWRRGSSAGGCRNHPGTFWTNPQFKISLPEGDDPEDDAEGNVVACTCLVALMQKNWRHARQQGAQLQTIGFVLYAVPKEFQNIQDVHLKKEFFTKYQDHGFSEIFTNSREVSSQLRLPPGEYIIIPSTFEPHRDADFLLRVFTEKHSESWELDEVNYAEQLQEEKVSEDDMDQDFLHLFKIVAGEGKEIGVYELQRLLNRMAIKFKSFKTKGFGLDACRCMINLMDKDGSGKLGLLEFKILWKKLKKWMDIFRECDQDHSGTLNSYEMRLVIEKAGIKLNNKVMQVLVARYADDDLIIDFDSFISCFLRLKTMFTFFLTMDPKNTGHICLSLEQWLQMTMWG | |
RUO | |
CAPN11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
11131 | |
CAPN11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
calpain11 | |
CAPN11 | |
Recombinant | |
wheat germ expression system |