missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cadherin-9 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
261.00€ - 549.00€
Specifications
| Antigen | Cadherin-9 |
|---|---|
| Clone | 1087334 |
| Dilution | Immunohistochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30241019
|
R&D Systems
MAB11589-SP |
25 μg |
261.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30241012
|
R&D Systems
MAB11589-100 |
100 μg |
549.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadherin-9 Monoclonal antibody specifically detects Cadherin-9 in Human samples. It is validated for ImmunohistochemistrySpecifications
| Cadherin-9 | |
| Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| cadherin 9, type 2 (T1-cadherin), Cadherin9, CDH9, T1-Cadherin | |
| Human embryonic kidney cell, HEK293-derived human Cadherin-9, Gly54-Ala615, Accession # Q9ULB4 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1087334 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9ULB4 | |
| 1007 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title