missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C1orf25 Full-length ORF (AAH45535.1, 1 a.a. - 733 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00081627-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16165193

  • 335.00€ / 10µg
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein that has some similarity to N2,N2-dimethylguanosine tRNA methyltransferase from other organisms. Studies of the mouse ortholog have shown that this protein plays a role in motor coordination and exploratory behavior, and it may also be involved in modulating postnatal neuronal functions. Alternatively spliced transcripts have been identified for this gene, but their full-length nature is not known. [provided by RefSeq]

Sequence: MENMAEEELLPLEKEEVEVAQVQVPTPARDSAGVPAPAPDSALDSAPTPASAPAPAPALAQAPALSPSLASAPEEAKSKRHISIQRQLADLENLAFVTDGNFDSASSLNSDNLDAGNRQACPLCPKEKFRACNSHKLRRHLQNLHWKVSVEFEGYRMCICHLPCRPVKPNIIGEQITSKMGAHYHCIICSATITRRTDMLGHVRRHMNKGETKSSYIAASTAKPPKEILKEADTDVQVCPNYSIPQKTDSYFNPKMKLNRQLIFCTLAALAEERKPLECLDAFGATGIMGLQWAKHLGNAVKVTINDLNENSVTLIQENCHLNKLKVVVDSKEKEKSDDILEEGEKNLGNIKVTKMDANVLMHLRSFDFIHLDPFGTSVNYLDSAFRNIRNLGIVSVTSTDISSLYAKAQHVARRHYGCNIVRTEYYKELAARIVVAAVARAAARCNKGIEVLFAVALEHFVLVVVRVLRGPTSADETAKKIQYLIHCQWCEERIFQKDGNMVEENPYRQLPCNCHGSMPGKTAIELGPLWSSSLFNTGFLKRMLFESLHHGLDDIQTLIKTLIFESECTPQSQFSIHASSNVNKQEENGVFIKTTDDTTTDNYIAQGKRKSNEMITNLGKKQKTDVSTEHPPFYYNIHRHSIKGMNMPKLKKFLCYLSQAGFRVSRTHFDPMGVRTDAPLMQFKSILLKYSTPTYTGGQSESHVQSASEDTVTERVEMSVNDKAEASGCRRW
Specifications

Specifications

AAH45535.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
108.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC25112/MGC57134/MST070/MSTP070/TRM1L/bG120K12.3
C1orf25
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
81627
C1orf25 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MENMAEEELLPLEKEEVEVAQVQVPTPARDSAGVPAPAPDSALDSAPTPASAPAPAPALAQAPALSPSLASAPEEAKSKRHISIQRQLADLENLAFVTDGNFDSASSLNSDNLDAGNRQACPLCPKEKFRACNSHKLRRHLQNLHWKVSVEFEGYRMCICHLPCRPVKPNIIGEQITSKMGAHYHCIICSATITRRTDMLGHVRRHMNKGETKSSYIAASTAKPPKEILKEADTDVQVCPNYSIPQKTDSYFNPKMKLNRQLIFCTLAALAEERKPLECLDAFGATGIMGLQWAKHLGNAVKVTINDLNENSVTLIQENCHLNKLKVVVDSKEKEKSDDILEEGEKNLGNIKVTKMDANVLMHLRSFDFIHLDPFGTSVNYLDSAFRNIRNLGIVSVTSTDISSLYAKAQHVARRHYGCNIVRTEYYKELAARIVVAAVARAAARCNKGIEVLFAVALEHFVLVVVRVLRGPTSADETAKKIQYLIHCQWCEERIFQKDGNMVEENPYRQLPCNCHGSMPGKTAIELGPLWSSSLFNTGFLKRMLFESLHHGLDDIQTLIKTLIFESECTPQSQFSIHASSNVNKQEENGVFIKTTDDTTTDNYIAQGKRKSNEMITNLGKKQKTDVSTEHPPFYYNIHRHSIKGMNMPKLKKFLCYLSQAGFRVSRTHFDPMGVRTDAPLMQFKSILLKYSTPTYTGGQSESHVQSASEDTVTERVEMSVNDKAEASGCRRW
RUO
C1orf25
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers