missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C1orf25 Full-length ORF (AAH45535.1, 1 a.a. - 733 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH45535.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 81627 |
Molecular Weight (g/mol) | 108.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16175193
|
Abnova™
H00081627-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16165193
|
Abnova™
H00081627-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein that has some similarity to N2,N2-dimethylguanosine tRNA methyltransferase from other organisms. Studies of the mouse ortholog have shown that this protein plays a role in motor coordination and exploratory behavior, and it may also be involved in modulating postnatal neuronal functions. Alternatively spliced transcripts have been identified for this gene, but their full-length nature is not known. [provided by RefSeq]
Sequence: MENMAEEELLPLEKEEVEVAQVQVPTPARDSAGVPAPAPDSALDSAPTPASAPAPAPALAQAPALSPSLASAPEEAKSKRHISIQRQLADLENLAFVTDGNFDSASSLNSDNLDAGNRQACPLCPKEKFRACNSHKLRRHLQNLHWKVSVEFEGYRMCICHLPCRPVKPNIIGEQITSKMGAHYHCIICSATITRRTDMLGHVRRHMNKGETKSSYIAASTAKPPKEILKEADTDVQVCPNYSIPQKTDSYFNPKMKLNRQLIFCTLAALAEERKPLECLDAFGATGIMGLQWAKHLGNAVKVTINDLNENSVTLIQENCHLNKLKVVVDSKEKEKSDDILEEGEKNLGNIKVTKMDANVLMHLRSFDFIHLDPFGTSVNYLDSAFRNIRNLGIVSVTSTDISSLYAKAQHVARRHYGCNIVRTEYYKELAARIVVAAVARAAARCNKGIEVLFAVALEHFVLVVVRVLRGPTSADETAKKIQYLIHCQWCEERIFQKDGNMVEENPYRQLPCNCHGSMPGKTAIELGPLWSSSLFNTGFLKRMLFESLHHGLDDIQTLIKTLIFESECTPQSQFSIHASSNVNKQEENGVFIKTTDDTTTDNYIAQGKRKSNEMITNLGKKQKTDVSTEHPPFYYNIHRHSIKGMNMPKLKKFLCYLSQAGFRVSRTHFDPMGVRTDAPLMQFKSILLKYSTPTYTGGQSESHVQSASEDTVTERVEMSVNDKAEASGCRRWSpecifications
AAH45535.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
108.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MENMAEEELLPLEKEEVEVAQVQVPTPARDSAGVPAPAPDSALDSAPTPASAPAPAPALAQAPALSPSLASAPEEAKSKRHISIQRQLADLENLAFVTDGNFDSASSLNSDNLDAGNRQACPLCPKEKFRACNSHKLRRHLQNLHWKVSVEFEGYRMCICHLPCRPVKPNIIGEQITSKMGAHYHCIICSATITRRTDMLGHVRRHMNKGETKSSYIAASTAKPPKEILKEADTDVQVCPNYSIPQKTDSYFNPKMKLNRQLIFCTLAALAEERKPLECLDAFGATGIMGLQWAKHLGNAVKVTINDLNENSVTLIQENCHLNKLKVVVDSKEKEKSDDILEEGEKNLGNIKVTKMDANVLMHLRSFDFIHLDPFGTSVNYLDSAFRNIRNLGIVSVTSTDISSLYAKAQHVARRHYGCNIVRTEYYKELAARIVVAAVARAAARCNKGIEVLFAVALEHFVLVVVRVLRGPTSADETAKKIQYLIHCQWCEERIFQKDGNMVEENPYRQLPCNCHGSMPGKTAIELGPLWSSSLFNTGFLKRMLFESLHHGLDDIQTLIKTLIFESECTPQSQFSIHASSNVNKQEENGVFIKTTDDTTTDNYIAQGKRKSNEMITNLGKKQKTDVSTEHPPFYYNIHRHSIKGMNMPKLKKFLCYLSQAGFRVSRTHFDPMGVRTDAPLMQFKSILLKYSTPTYTGGQSESHVQSASEDTVTERVEMSVNDKAEASGCRRW | |
RUO | |
C1orf25 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
81627 | |
C1orf25 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC25112/MGC57134/MST070/MSTP070/TRM1L/bG120K12.3 | |
C1orf25 | |
Recombinant | |
wheat germ expression system |