Learn More
Abnova™ Human C1orf25 Full-length ORF (AAH45535.1, 1 a.a. - 733 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00081627-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that has some similarity to N2,N2-dimethylguanosine tRNA methyltransferase from other organisms. Studies of the mouse ortholog have shown that this protein plays a role in motor coordination and exploratory behavior, and it may also be involved in modulating postnatal neuronal functions. Alternatively spliced transcripts have been identified for this gene, but their full-length nature is not known. [provided by RefSeq]
Sequence: MENMAEEELLPLEKEEVEVAQVQVPTPARDSAGVPAPAPDSALDSAPTPASAPAPAPALAQAPALSPSLASAPEEAKSKRHISIQRQLADLENLAFVTDGNFDSASSLNSDNLDAGNRQACPLCPKEKFRACNSHKLRRHLQNLHWKVSVEFEGYRMCICHLPCRPVKPNIIGEQITSKMGAHYHCIICSATITRRTDMLGHVRRHMNKGETKSSYIAASTAKPPKEILKEADTDVQVCPNYSIPQKTDSYFNPKMKLNRQLIFCTLAALAEERKPLECLDAFGATGIMGLQWAKHLGNAVKVTINDLNENSVTLIQENCHLNKLKVVVDSKEKEKSDDILEEGEKNLGNIKVTKMDANVLMHLRSFDFIHLDPFGTSVNYLDSAFRNIRNLGIVSVTSTDISSLYAKAQHVARRHYGCNIVRTEYYKELAARIVVAAVARAAARCNKGIEVLFAVALEHFVLVVVRVLRGPTSADETAKKIQYLIHCQWCEERIFQKDGNMVEENPYRQLPCNCHGSMPGKTAIELGPLWSSSLFNTGFLKRMLFESLHHGLDDIQTLIKTLIFESECTPQSQFSIHASSNVNKQEENGVFIKTTDDTTTDNYIAQGKRKSNEMITNLGKKQKTDVSTEHPPFYYNIHRHSIKGMNMPKLKKFLCYLSQAGFRVSRTHFDPMGVRTDAPLMQFKSILLKYSTPTYTGGQSESHVQSASEDTVTERVEMSVNDKAEASGCRRWSpecifications
AAH45535.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
108.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC25112/MGC57134/MST070/MSTP070/TRM1L/bG120K12.3 | |
C1orf25 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
81627 | |
C1orf25 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MENMAEEELLPLEKEEVEVAQVQVPTPARDSAGVPAPAPDSALDSAPTPASAPAPAPALAQAPALSPSLASAPEEAKSKRHISIQRQLADLENLAFVTDGNFDSASSLNSDNLDAGNRQACPLCPKEKFRACNSHKLRRHLQNLHWKVSVEFEGYRMCICHLPCRPVKPNIIGEQITSKMGAHYHCIICSATITRRTDMLGHVRRHMNKGETKSSYIAASTAKPPKEILKEADTDVQVCPNYSIPQKTDSYFNPKMKLNRQLIFCTLAALAEERKPLECLDAFGATGIMGLQWAKHLGNAVKVTINDLNENSVTLIQENCHLNKLKVVVDSKEKEKSDDILEEGEKNLGNIKVTKMDANVLMHLRSFDFIHLDPFGTSVNYLDSAFRNIRNLGIVSVTSTDISSLYAKAQHVARRHYGCNIVRTEYYKELAARIVVAAVARAAARCNKGIEVLFAVALEHFVLVVVRVLRGPTSADETAKKIQYLIHCQWCEERIFQKDGNMVEENPYRQLPCNCHGSMPGKTAIELGPLWSSSLFNTGFLKRMLFESLHHGLDDIQTLIKTLIFESECTPQSQFSIHASSNVNKQEENGVFIKTTDDTTTDNYIAQGKRKSNEMITNLGKKQKTDVSTEHPPFYYNIHRHSIKGMNMPKLKKFLCYLSQAGFRVSRTHFDPMGVRTDAPLMQFKSILLKYSTPTYTGGQSESHVQSASEDTVTERVEMSVNDKAEASGCRRW | |
RUO | |
C1orf25 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |