missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C1orf22 Full-length ORF (AAH16464.1, 1 a.a. - 122 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080267-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124893

  • 508.00€ / 25µg
Estimated Shipment: 07-05-2024
to see stock.
Add to basket

Description

Description

Quality control in the endoplasmic reticulum (ER) ensures that only properly folded proteins are retained in the cell through recognition and degradation of misfolded or unassembled proteins. EDEM3 belongs to a group of proteins that accelerate degradation of misfolded glycoproteins in the ER (Hirao et al., 2006 [PubMed 16431915]).[supplied by OMIM]

Sequence: MGGFKYGDEQPRSDWRSYRRNLEHAVLELTLFKTVPSKMEIHSSPFKCSTAPPCNTSGQGKITEHSCEPDFCCLWIDKKQNSFSSGVGNRSLDSLLIKGSSPFLVLGVRGSFGKMHPSIVAF
Specifications

Specifications

AAH16464.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1orf22
EDEM3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
80267
C1orf22 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGGFKYGDEQPRSDWRSYRRNLEHAVLELTLFKTVPSKMEIHSSPFKCSTAPPCNTSGQGKITEHSCEPDFCCLWIDKKQNSFSSGVGNRSLDSLLIKGSSPFLVLGVRGSFGKMHPSIVAF
RUO
EDEM3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers