missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C1orf22 Full-length ORF (AAH16464.1, 1 a.a. - 122 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH16464.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80267 |
Molecular Weight (g/mol) | 39.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124893
|
Abnova™
H00080267-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16114893
|
Abnova™
H00080267-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
Quality control in the endoplasmic reticulum (ER) ensures that only properly folded proteins are retained in the cell through recognition and degradation of misfolded or unassembled proteins. EDEM3 belongs to a group of proteins that accelerate degradation of misfolded glycoproteins in the ER (Hirao et al., 2006 [PubMed 16431915]).[supplied by OMIM]
Sequence: MGGFKYGDEQPRSDWRSYRRNLEHAVLELTLFKTVPSKMEIHSSPFKCSTAPPCNTSGQGKITEHSCEPDFCCLWIDKKQNSFSSGVGNRSLDSLLIKGSSPFLVLGVRGSFGKMHPSIVAFSpecifications
AAH16464.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGGFKYGDEQPRSDWRSYRRNLEHAVLELTLFKTVPSKMEIHSSPFKCSTAPPCNTSGQGKITEHSCEPDFCCLWIDKKQNSFSSGVGNRSLDSLLIKGSSPFLVLGVRGSFGKMHPSIVAF | |
RUO | |
EDEM3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
80267 | |
C1orf22 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C1orf22 | |
EDEM3 | |
Recombinant | |
wheat germ expression system |