missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTLA Antibody, R&D Systems™

Rabbit Monoclonal Antibody
195.00€ - 457.00€
Specifications
| Antigen | BTLA/CD272 |
|---|---|
| Clone | 2161A |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792217
![]() |
R&D Systems
MAB33542-SP |
25 μg |
195.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18714527
![]() |
R&D Systems
MAB33542-100 |
100 μg |
457.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| BTLA/CD272 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
| Mouse myeloma cell line, NS0-derived human BTLA, Ile25-Ser150, Accession # AAP44003 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human BTLA in direct ELISAs. |
| 2161A | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AAP44003 | |
| 151888 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title