missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTK Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB11593-100
This item is not returnable.
View return policy
Description
BTK Monoclonal antibody specifically detects BTK in Human samples. It is validated for Western Blot
Specifications
| BTK | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| Agammaglobulinaemia tyrosine kinase, AGMX1, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, IMD1, PSCTK1, tyrosine-protein kinase BTK, XLA, XLAMGC126261 | |
| E. coli -derived recombinant human BTK, Glu280-Val377, Accession # Q06187 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 3005A | |
| Western Blot 2 μg/mL | |
| Q06187 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 695 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction