Learn More
Abnova™ Human BIRC4BP Partial ORF (NP_059993.2, 202 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054739-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
X-linked inhibitor of apoptosis (XIAP; MIM 300079) is a potent member of the IAP family. All members of this family possess baculoviral IAP (BIR) repeats, cysteine-rich domains of approximately 80 amino acids that bind and inhibit caspases (e.g., CASP3; MIM 600636). XIAP has 3 BIR domains and a C-terminal RING zinc finger that possesses E3 ubiquitin ligase (see MIM 601623) activity. XAF1 antagonizes the anticaspase activity of XIAP and may be important in mediating apoptosis resistance in cancer cells (Liston et al., 2001 [PubMed 11175744]).[supplied by OMIM]
Sequence: QTSTMEKDVRPKTRSINRFPLHSESSSKKAPRSKNKTLDPLLMSEPKPRTSSPRGDKAAYDILRRCSQCGILLPLPILNQHQEKCRWLASSKGKQVRNFSSpecifications
NP_059993.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QTSTMEKDVRPKTRSINRFPLHSESSSKKAPRSKNKTLDPLLMSEPKPRTSSPRGDKAAYDILRRCSQCGILLPLPILNQHQEKCRWLASSKGKQVRNFS | |
RUO | |
XAF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
54739 | |
BIRC4BP (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIRC4BP/HSXIAPAF1/XIAPAF1 | |
XAF1 | |
Recombinant | |
wheat germ expression system |