Learn More
Abnova™ Human BIRC4BP Partial ORF (NP_059993.2, 202 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_059993.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54739 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16147716
|
Abnova™
H00054739-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16157716
|
Abnova™
H00054739-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
X-linked inhibitor of apoptosis (XIAP; MIM 300079) is a potent member of the IAP family. All members of this family possess baculoviral IAP (BIR) repeats, cysteine-rich domains of approximately 80 amino acids that bind and inhibit caspases (e.g., CASP3; MIM 600636). XIAP has 3 BIR domains and a C-terminal RING zinc finger that possesses E3 ubiquitin ligase (see MIM 601623) activity. XAF1 antagonizes the anticaspase activity of XIAP and may be important in mediating apoptosis resistance in cancer cells (Liston et al., 2001 [PubMed 11175744]).[supplied by OMIM]
Sequence: QTSTMEKDVRPKTRSINRFPLHSESSSKKAPRSKNKTLDPLLMSEPKPRTSSPRGDKAAYDILRRCSQCGILLPLPILNQHQEKCRWLASSKGKQVRNFSSpecifications
NP_059993.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIRC4BP/HSXIAPAF1/XIAPAF1 | |
XAF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54739 | |
BIRC4BP (Human) Recombinant Protein (Q01) | |
QTSTMEKDVRPKTRSINRFPLHSESSSKKAPRSKNKTLDPLLMSEPKPRTSSPRGDKAAYDILRRCSQCGILLPLPILNQHQEKCRWLASSKGKQVRNFS | |
RUO | |
XAF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |