missing translation for 'onlineSavingsMsg'
Learn More

Human beta-1,4-Galactosyltransferase 1/B4GalT1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18786968
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18786968 25 μg 25µg
18708649 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18786968 Supplier R&D Systems Supplier No. MAB36091SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-1,4-Galactosyltransferase 1/B4GalT1 Monoclonal specifically detects beta-1,4-Galactosyltransferase 1/B4GalT1 in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-1,4-Galactosyltransferase 1/B4GalT1
Applications Immunocytochemistry
Classification Monoclonal
Clone 394709
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P15291
Gene Alias 1,4- galactosyltransferase, polypeptide 1, B4GAL-T1, beta-1,4-galactosyltransferase 1, Beta-1,4-GalTase 1, Beta4Gal-T1, betaGlcNAc beta, CDG2D, EC 2.4.1, GGTB2, GGTB2DKFZp686N19253, glycoprotein-4-beta-galactosyltransferase 2, GT1, GTB, lactose synthase, MGC50983, UDP-Gal, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 1, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human beta-1,4-Galactosyltransferase 1/B4GalT1 Ala44-Ser398 Accession # P15291
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 2683
Test Specificity Detects human beta-1,4-Galactosyltransferase 1/B4GalT1in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.