Learn More
Human BATF3 Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  R&D Systems AF7437
Product Code. 18089541
  • 615.00€ / 100µg
Description
BATF3 Polyclonal specifically detects BATF3 in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BATF3
Polyclonal
Unconjugated
Q9NR55
BATF3
E. coli-derived recombinant human BATF3 Met1-Arg127 Accession # Q9NR55
100 μg
Primary
Detects human BATF3 in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse BATF3 is observed, and less than 2% cross-reactivity with recombinant human BATF is observed.
Human
IgG
Western Blot, Immunocytochemistry
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT, SNFT21-kD small nuclear factor isolated from T cells
Sheep
Affinity Purified
RUO
55509
Sterile PBS to a final concentration of 0.2 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.