missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ASCL2/Mash2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6539
This item is not returnable.
View return policy
Description
ASCL2/Mash2 Polyclonal specifically detects ASCL2/Mash2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ASCL2/Mash2 | |
| Polyclonal | |
| Unconjugated | |
| Q99929 | |
| ASCL2 | |
| E. coli-derived recombinant human ASCL2/Mash2 Met1-Ala49 Accession # Q99929 | |
| 100 μg | |
| Primary | |
| Detects human ASCL2/Mash2 in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| achaete-scute complex (Drosophila) homolog-like 2, achaete-scute complex homolog 2 (Drosophila), achaete-scute complex-like 2 (Drosophila), ASH2, ASH-2, BHLHA45, bHLHa45achaete-scute homolog 2, Class A basic helix-loop-helix protein 45, HASH2, HASH2achaete-scute complex-like 2, mammalian achaete/scute homologue 2, MASH2 | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| 430 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction