missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aquaporin 4/AQP4 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11659-100
This item is not returnable.
View return policy
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Multiplex Immunofluorescence,Immunohistochemistry
Specifications
| Aquaporin-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS. | |
| AQP4, AQP-4, aquaporin 4, MGC22454, MIWC, WCH4 | |
| Synthetic Peptide, Accession # P55087 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Multiplex Immunofluorescence, Immunohistochemistry | |
| 1093722 | |
| Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 25 μg/mL | |
| P55087 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 361 | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction