missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AOC2 Partial ORF (NP_033720, 169 a.a. - 268 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000314-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16164704

  • 545.00€ / 25µg

Estimated Shipment: 02-08-2022
to see stock.
Add to basket

Description

Description

Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes and ammonia in the presence of copper and quinone cofactor. This gene shows high sequence similarity to copper amine oxidases from various species ranging from bacteria to mammals. The protein contains several conserved motifs including the active site of amine oxidases and the histidine residues that likely bind copper. It may be a critical modulator of signal transmission in retina, possibly by degrading the biogenic amines dopamine, histamine, and putrescine. This gene may be a candidate gene for hereditary ocular diseases. Alternate splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: RAEFTQMWRHLKEVELPKAPIFLSSTFNYNGSTLAAVHATPRGLRSGDRATWMALYHNISGVGLFLHPVGLELLLDHRALDPAHWTVQQVFYLGHYYADL
Specifications

Specifications

NP_033720
Liquid
314
AOC2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DAO2/RAO
AOC2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RAEFTQMWRHLKEVELPKAPIFLSSTFNYNGSTLAAVHATPRGLRSGDRATWMALYHNISGVGLFLHPVGLELLLDHRALDPAHWTVQQVFYLGHYYADL
RUO
AOC2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers