Learn More
Abnova™ Human AOC2 Partial ORF (NP_033720, 169 a.a. - 268 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_033720 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 314 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16164704
|
Abnova™
H00000314-Q01L |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16154704
|
Abnova™
H00000314-Q01S |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes and ammonia in the presence of copper and quinone cofactor. This gene shows high sequence similarity to copper amine oxidases from various species ranging from bacteria to mammals. The protein contains several conserved motifs including the active site of amine oxidases and the histidine residues that likely bind copper. It may be a critical modulator of signal transmission in retina, possibly by degrading the biogenic amines dopamine, histamine, and putrescine. This gene may be a candidate gene for hereditary ocular diseases. Alternate splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: RAEFTQMWRHLKEVELPKAPIFLSSTFNYNGSTLAAVHATPRGLRSGDRATWMALYHNISGVGLFLHPVGLELLLDHRALDPAHWTVQQVFYLGHYYADLSpecifications
NP_033720 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAO2/RAO | |
AOC2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
314 | |
AOC2 (Human) Recombinant Protein (Q01) | |
RAEFTQMWRHLKEVELPKAPIFLSSTFNYNGSTLAAVHATPRGLRSGDRATWMALYHNISGVGLFLHPVGLELLLDHRALDPAHWTVQQVFYLGHYYADL | |
RUO | |
AOC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |