missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
208.00€ - 2020.00€
Specifications
| Antigen | Angiopoietin-2 |
|---|---|
| Clone | 180102 |
| Concentration | LYOPH |
| Dilution | Western Blot 0.5 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18761049
![]() |
R&D Systems
MAB0983-SP |
25 μg |
208.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744428
![]() |
R&D Systems
MAB0983-100 |
100 μg |
502.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18736929
![]() |
R&D Systems
MAB0983-500 |
500 μg |
2020.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human samples. It is validated for Western Blot.Specifications
| Angiopoietin-2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, ANGPT2, Tie2-ligand | |
| ANGPT2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 180102 | |
| Western Blot 0.5 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| O15123 | |
| 285 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-2 Asp68-Phe496 Accession # O15123 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Angiopoietin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Ang-1, rhAng-4, rhAng-X, or rmANGPTL3. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title