missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ALDH18A1 Partial ORF (NP_002851, 696 a.a. - 795 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005832-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16113975

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene. [provided by RefSeq]

Sequence: TDVIVTEDENTAEFFLQHVDSACVFWNASTRFSDGYRFGLGAEVGISTSRIHARGPVGLEGLLTTKWLLRGKDHVVSDFSEHGSLKYLHENLPIPQRNTN
Specifications

Specifications

NP_002851
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TDVIVTEDENTAEFFLQHVDSACVFWNASTRFSDGYRFGLGAEVGISTSRIHARGPVGLEGLLTTKWLLRGKDHVVSDFSEHGSLKYLHENLPIPQRNTN
RUO
ALDH18A1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5832
ALDH18A1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GSAS/MGC117316/P5CS/PYCS
ALDH18A1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers