Learn More
Abnova™ Human ALDH18A1 Partial ORF (NP_002851, 696 a.a. - 795 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_002851 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5832 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16123975
|
Abnova™
H00005832-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16113975
|
Abnova™
H00005832-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene. [provided by RefSeq]
Sequence: TDVIVTEDENTAEFFLQHVDSACVFWNASTRFSDGYRFGLGAEVGISTSRIHARGPVGLEGLLTTKWLLRGKDHVVSDFSEHGSLKYLHENLPIPQRNTNSpecifications
NP_002851 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GSAS/MGC117316/P5CS/PYCS | |
ALDH18A1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5832 | |
ALDH18A1 (Human) Recombinant Protein (Q01) | |
TDVIVTEDENTAEFFLQHVDSACVFWNASTRFSDGYRFGLGAEVGISTSRIHARGPVGLEGLLTTKWLLRGKDHVVSDFSEHGSLKYLHENLPIPQRNTN | |
RUO | |
ALDH18A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |