Learn More
Human Akt Pan Specific Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Brand: R&D Systems AF2055-SP
This item is not returnable.
View return policy
Description
AKT Polyclonal specifically detects AKT in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| AKT | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB alpha, PKB beta, PKB gamma, PKB-ALPHA, PRKBA, RAC, RAC-alpha, RAC-beta, RAC-gamma | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 207 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 20 ug/mL | |
| P31749 | |
| AKT1 | |
| Akt synthetic peptide IRCLQWTTVIERTFHV Accession # P31749 | |
| 25 μg | |
| Primary | |
| Detects human Akt1, Akt2, and Akt3 in Western blots. | |
| Human | |
| IgG |