Learn More
Abnova™ Human ADPRHL1 Full-length ORF (NP_612439.2, 1 a.a. - 354 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00113622-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ADP-ribosylation is a reversible posttranslational modification used to regulate protein function. ADP-ribosyltransferases (see ART1; MIM 601625) transfer ADP-ribose from NAD+ to the target protein, and ADP-ribosylhydrolases, such as ADPRHL1, reverse the reaction (Glowacki et al., 2002 [PubMed 12070318]).[supplied by OMIM]
Sequence: MEKFKAAMLLGSVGDALGYRNVCKENSTVGMKIQEELQRSGGLDHLVLSPGEWPVSDNTIMHIATAEALTTDYWCLDDLYREMVRCYVEIVEKLPERRPDPATIEGCAQLKPNNYLLAWHTPFNEKGSGFGAATKAMCIGLRYWKPERLETLIEVSVECGRMTHNHPTGFLGSLCTALFVSFAAQGKPLVQWGRDMLRAVPLAEEYCRKTIRHTAEYQEHWFYFEAKWQFYLEERKISKDSENKAIFPDNYDAEEREKTYRKWSSEGRGGRRGHDAPMIAYDALLAAGNSWTELCHRAMFHGGESAATGTIAGCLFGLLYGLDLVPKGLYQDLEDKEKLEDLGAALYRLSTEEKSpecifications
NP_612439.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ADPRHL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
113622 | |
ADPRHL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARH2 | |
ADPRHL1 | |
Recombinant | |
wheat germ expression system |