missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ADPRHL1 Full-length ORF (NP_612439.2, 1 a.a. - 354 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_612439.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 113622 |
Molecular Weight (g/mol) | 66.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16157013
|
Abnova™
H00113622-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16167013
|
Abnova™
H00113622-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
ADP-ribosylation is a reversible posttranslational modification used to regulate protein function. ADP-ribosyltransferases (see ART1; MIM 601625) transfer ADP-ribose from NAD+ to the target protein, and ADP-ribosylhydrolases, such as ADPRHL1, reverse the reaction (Glowacki et al., 2002 [PubMed 12070318]).[supplied by OMIM]
Sequence: MEKFKAAMLLGSVGDALGYRNVCKENSTVGMKIQEELQRSGGLDHLVLSPGEWPVSDNTIMHIATAEALTTDYWCLDDLYREMVRCYVEIVEKLPERRPDPATIEGCAQLKPNNYLLAWHTPFNEKGSGFGAATKAMCIGLRYWKPERLETLIEVSVECGRMTHNHPTGFLGSLCTALFVSFAAQGKPLVQWGRDMLRAVPLAEEYCRKTIRHTAEYQEHWFYFEAKWQFYLEERKISKDSENKAIFPDNYDAEEREKTYRKWSSEGRGGRRGHDAPMIAYDALLAAGNSWTELCHRAMFHGGESAATGTIAGCLFGLLYGLDLVPKGLYQDLEDKEKLEDLGAALYRLSTEEKSpecifications
NP_612439.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARH2 | |
ADPRHL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
113622 | |
ADPRHL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ADPRHL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |