missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ABCB6 Partial ORF (NP_005680, 743 a.a. - 842 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010058-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101766

  • 335.00€ / 10µg

Estimated Shipment: 27-04-2023
to see stock.
Add to basket

Description

Description

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This half-transporter likely plays a role in mitochondrial function. Localized to 2q26, this gene is considered a candidate gene for lethal neonatal metabolic syndrome, a disorder of mitochondrial function. [provided by RefSeq]

Sequence: KAPGIILLDEATSALDTSNERAIQASLAKVCANRTTIVVAHRLSTVVNADQILVIKDGCIVERGRHEALLSRGGVYADMWQLQQGQEETSEDTKPQTMER
Specifications

Specifications

NP_005680
Liquid
10058
ABCB6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ABC/ABC14/EST45597/FLJ22414/MTABC3/PRP/umat
ABCB6
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KAPGIILLDEATSALDTSNERAIQASLAKVCANRTTIVVAHRLSTVVNADQILVIKDGCIVERGRHEALLSRGGVYADMWQLQQGQEETSEDTKPQTMER
RUO
ABCB6
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers