Learn More
Invitrogen™ VSIG4 Monoclonal Antibody (JAV4), Brilliant Violet™ 605, eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 406-5757-42
Description
Description This JAV4 monoclonal antibody recognizes human V-Set and Immunoglobulin domain containing 4 (VSIG4), also known as Complement Receptor of the Immunoglobulin superfamily (CRIg) or Z39Ig. There are two variants of human VSIG4 that result from alternative splicing. Isoform 1 is longer and more prevalent than isoform 2, and this JAV4 antibody recognizes both. VSIG4 is a type I transmembrane glycoprotein structurally related to the B7 family of immune regulatory proteins. It contains one complete V-type Ig domain and one truncated C-type Ig domain. The JAV4 antibody will recognize VSIG4 on cells that have been formaldehyde-fixed and permeabilized. This antibody does not cross-react with mouse or rat VSIG4. Applications Tested This JAV4 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Blocking Buffers When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401</a>) or Brilliant Stain Buffer (Product # 00-4409-75...
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
Specifications
| VSIG4 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q9Y279 | |
| VSIG4 | |
| Fc-Protein and CHO cells. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| JAV4 | |
| Brilliant Violet 605 | |
| VSIG4 | |
| A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11326 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.