missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTRA1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29618-1-AP-150UL
This item is not returnable.
View return policy
Description
The HTRA1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human HTRA1. This antibody recognizes human antigen. The HTRA1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| HTRA1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5654 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 550 μg/mL | |
| ARMD7; HtrA; L56; ORF480; PRSS11 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 51 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction