missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR4 Monoclonal Antibody (631769)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561684
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
This gene is a member of the family of serotonin receptors, which are G protein coupled receptors that stimulate cAMP production in response to serotonin (5-hydroxytryptamine). The gene product is a glycosylated transmembrane protein that functions in both the peripheral and central nervous system to modulate the release of various neurotransmitters. Multiple transcript variants encoding proteins with distinct C-terminal sequences have been described, but the full-length nature of some transcript variants has not been determined.
Specifications
| HTR4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q13639 | |
| HTR4 | |
| NS0 mouse myeloma cell line transfected with human 5-HT4. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 631769 | |
| Unconjugated | |
| HTR4 | |
| 5 hydroxytryptamine ( serotonin) receptor 4; 5 hydroxytryptamine (serotonin) receptor 4; 5-HT>4L>; 5HT4; 5-HT4; 5-HT-4; 5HT4 receptor; 5-HT4 receptor; 5-HT4 receptor (two splicing variants L and S); 5-HT4R; 5HTR4; 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled; 5-hydroxytryptamine receptor 4; cardiac 5-HT4 receptor; Htr4; Serotonin receptor 4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3360 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction