missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPC111 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35414-20ul
This item is not returnable.
View return policy
Description
HSPC111 Polyclonal antibody specifically detects HSPC111 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| HSPC111 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| HBV pre-S2 trans-regulated protein 3, HSPC111HSPC185, LOC51491, NOP15, NOP16 nucleolar protein homolog (yeast), nucleolar protein 16, nucleolar protein 16 homolog, nucleolar protein 16 homolog (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-178 of human HSPC111 (NP_057475.2).,, Sequence:, MPKAKGKTRRQKFGYSVNRKRLNRNARRKAAPRIECSHIRHAWDHAKSVRQNLAEMGLAVDPNRAVPLRKRKVKAMEVDIEERPKELVRKPYVLNDLEAEASLPEKKGNTLSRDLIDYVRYMVENHGEDYKAMARDEKNYYQDTPKQIRSKINVYKRFYPAEWQDFLDSLQKRKMEVE | |
| 20 μL | |
| Stem Cell Markers | |
| 51491 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction