missing translation for 'onlineSavingsMsg'
Learn More

HspBAP1 Antibody (1A9), Novus Biologicals™

Product Code. 18479448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18479448 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18479448 Supplier Novus Biologicals Supplier No. H00079663M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HspBAP1 Monoclonal antibody specifically detects HspBAP1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HspBAP1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1A9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH11897
Gene Alias FLJ22623, heat shock 27 kDa associated protein, HSPB (heat shock 27kD) associated protein 1, HSPB (heat shock 27kDa) associated protein 1, HSPB1-associated protein 1,27 kDa heat shock protein-associated protein 1, PASS1FLJ39386, Protein associated with small stress protein 1, protein associating with small stress protein PASS1
Host Species Mouse
Immunogen HSPBAP1 (AAH11897, 1 a.a. ∽ 488 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAGSEATTPVIVAAGAGGEEGEHVKPFKPEKAKEIIMSLQQPAIFCNMVFDWPARHWNAKYLSQVLHGKQIRFRMGMKSMSTVPQFETTCNYVEATLEEFLTWNCDQSSISGPFRDYDHSKFWAYADYKYFVSLFEDKTDLFQDVKWSDFGFPGRNGQESTLWIGSLGAHTPCHLDSYGCNLVFQVQGRKRWHLFPPEDTPFLYPTRIPYEESSVFSKINVVNPDLKRFPQFRKAQRHAVTLSPGQVLFVPRHWWHYVESIDPVTVSINSWIELEEDHLARVEEAITRMLVCALKTAENPQNTRAWLNPTEVEETSHAVNCCYLNAAVSAFFDRCRTSEVVEIQALRTDGEHMKKEELNVCNHMEVGQTGSQNLTTGTDKPEAASPFGPDLVPVAQRSEEPPSERGGIFGSDGKDFVDKDGGHFGKLHCAKRQQIMSNSENAIEEQIASNTTTTPQTFISTDDLLDCLVNPQVTRIVAQLLIQGRSL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79663
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.